Boron nitride 31% refractory brushable paint

Supplier: Thermo Scientific Chemicals
Warning

44086-18 44086-30 44086-A1
CAAA44086-18EA 82.79 CAD
CAAA44086-18 CAAA44086-30 CAAA44086-A1
Boron nitride 31% refractory brushable paint
Boron nitride

Ready to use for thinner coating. Dry coating contains 94% BN


CAS No.: 10043-11-5 Molecular Formula: BN Physical Form: Paste Appearance: White Odor: Odorless MDL No.: MFCD00011317

Formula: BN
MW: 24.82 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011317
CAS Number: 10043-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR