Poly(vinylidene fluoride)
Supplier: Thermo Scientific Chemicals
Poly(vinylidene fluoride) is useful in coatings, film, filter cloth, instrument linings, filtration membranes, pump parts, linings and valves. It is also employed as a material for transducers in devices such as headphones, microphones and sonic detectors. It is also essential for piezoelectric and electrostrictive applications. In addition, it is useful for monofilament fishing lines replacing nylon monofilament. It acts as a standard binder used in the production of composite electrodes for lithium ion batteries. It serves as a high grade insulator for wires, nanomaterials and pressure sensors. It is utilized to make filters which are used to prepare samples for high performance liquid chromatography and other advanced analytical techniques.
Notes: Incompatible with strong oxidizing agents.
CAS No.: 24937-79-9
Molecular Formula: (-CH2CF2-)n
Physical Form: Powder
Appearance: White
Odor: Odorless
Melting Point: 155-160°
MDL No.: MFCD00084470
Formula:
(-CH₂CF₂-)n Melting Pt: 155…160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00084470 CAS Number: 24937-79-9 |
Specification Test Results
Appearance (Color) | White |
Form | Powder |
Viscosity | Melt viscosity 23.50 to 29.50 Kpoise |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...