Zinc copper composite ≥99% (metals basis) -100 mesh

Supplier: Thermo Scientific Chemicals
Warning

44013-09 44013-22 44013-36
CAAA44013-09EA 45.35 CAD
CAAA44013-09 CAAA44013-22 CAAA44013-36
Zinc copper composite ≥99% (metals basis) -100 mesh
Zinc copper composite

Purity: 99% Molecular Formula: Typically Zn 90-95, Cu 5-10 wt% Physical Form: Alloy Precursor UN No.: 3089 MDL No.: MFCD03102068

Formula: ∼Zn 90-95,Cu 5-10%
Storage Temperature: Ambient
MDL Number: MFCD03102068
UN: 3089
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR