Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')cobalt(III) 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(III), Co(TMHD)3

43993-02 43993-04 43993-09
CAAA43993-02EA 102.38 CAD
CAAA43993-02 CAAA43993-04 CAAA43993-09
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')cobalt(III) 99%
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')cobalt(III)

Applications: Vapor deposition


Purity: 99% CAS No.: 14877-41-9 Molecular Formula: C33H57CoO6 Formula Weight: 608.72 Physical Form: Powder Appearance: Green Melting Point: 254-256° Boiling Point: subl. 120°/0.5mm MDL No.: MFCD00233617


Vapor deposition

Formula: C₃₃H₅₇CoO₆
MW: 608.735 g/mol
Boiling Pt: 120 °C (0.5 mmHg) (subl.)
Melting Pt: 254…256 °C
Storage Temperature: Ambient
MDL Number: MFCD00233617
CAS Number: 14877-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR