Poly(methyl methacrylate)

Supplier: Thermo Scientific Chemicals

43982-18 43982-30 43982-A1
CAAA43982-18EA 65.41 CAD
CAAA43982-18 CAAA43982-30 CAAA43982-A1
Poly(methyl methacrylate)
Poly(methyl methacrylate)

Average MW 350,000


CAS No.: 9011-14-7 Molecular Formula: [-CH2C(CH3)(CO2CH3)-]n Physical Form: Powder Appearance: White Melting Point: 150° MDL No.: MFCD00134349

Formula: C₅H₈O₂([-CH₂C(CH₃)(CO₂CH₃)-]n)
Melting Pt: >150 °C
Storage Temperature: Ambient
MDL Number: MFCD00134349
CAS Number: 9011-14-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR