Magnesium oxalate dihydrate ≥98.5%

Supplier: Thermo Scientific Chemicals
Warning

43976-22 43976-36 43976-A3
CAAA43976-22EA 101.44 CAD
CAAA43976-22 CAAA43976-36 CAAA43976-A3
Magnesium oxalate dihydrate ≥98.5%
Magnesium oxalate dihydrate

Purity: 98.5+% CAS No.: 547-66-0 Molecular Formula: MgC2O4.2H2O Formula Weight: 148.36 (112.32anhy) Physical Form: Powder/Pieces Odor: Odorless Merck Reference: 14,5676 UN No.: 3288 MDL No.: MFCD00287279

Formula: C₂MgO₄·2H₂O
MW: 148.36 g/mol (112.32 anhy)
Melting Pt: 150 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00287279
CAS Number: 547-66-0
EINECS: 208-932-1
UN: 3288
ADR: 6.1,III
Merck Index: 14,5676

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR