Silver Protein strong, powder for histology

Supplier: Thermo Scientific Chemicals

43970-06 43970-14 43970-22
CAAA43970-06EA 56.56 CAD
CAAA43970-06 CAAA43970-14 CAAA43970-22
Silver Protein strong, powder for histology
Silver protein

Silver protein is used in electron microscopy with periodic acid and thiocarbohydrazide or thiosemicarbohydrazide as a positive stain for carbohydrates such as glycogen. It can also be used for light microscopy to stain nerve tissue. It is also used in histological stain for microscopic analysis.


  • Silver content: min 7.5%
  • Hygroscopic
  • Light Sensitive
  • Store under Nitrogen
  • Soluble in water. Slightly soluble in methanol, diethyl ether


CAS No.: 9008-42-8 Physical Form: Powder Sensitivity: Light Sensitive; Hygroscopic Appearance: Pale yellow to brown MDL No.: MFCD00132268


Applications: Histological stain for microscopic analysis

Formula: Ag (with protein)
Storage Temperature: Ambient
MDL Number: MFCD00132268
CAS Number: 9008-42-8
Merck Index: 14,8527

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR