Poly(methyl methacrylate) 600 micron

Supplier: Thermo Scientific Chemicals

43969-18 43969-30
CAAA43969-18EA 156.26 CAD
CAAA43969-18 CAAA43969-30
Poly(methyl methacrylate) 600 micron
Poly(methyl methacrylate)

Average MW 120,000


CAS No.: 9011-14-7 Molecular Formula: [-CH2C(CH3)(CO2CH3)-]n Physical Form: Powder MDL No.: MFCD00134349

Formula: C₅H₈O₂([-CH₂C(CH₃)(CO₂CH₃)-]n)
Storage Temperature: Ambient
MDL Number: MFCD00134349
CAS Number: 9011-14-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR