Polyethylene UHMW

Supplier: Thermo Scientific Chemicals

43951-18 43951-30 43951-A1
CAAA43951-18EA 111.71 CAD
CAAA43951-18 CAAA43951-30 CAAA43951-A1
Polyethylene UHMW
Polyethylene (CAS 9002-88-4)

CAS No.: 9002-88-4 Molecular Formula: (CH2CH2)n Physical Form: Powder Odor: Odorless MDL No.: MFCD00084423

Formula: (CH₂CH₂)n
Storage Temperature: Ambient
MDL Number: MFCD00084423
CAS Number: 9002-88-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR