Cobalt(II) sulfamate hydrate

Supplier: Thermo Scientific Chemicals
Warning

43906-22 43906-36 43906-A3
CAAA43906-22EA 206.98 CAD
CAAA43906-22 CAAA43906-36 CAAA43906-A3
Cobalt(II) sulfamate hydrate
Cobalt(II) sulfamate hydrate

CAS No.: 14017-41-5 Molecular Formula: Co(SO3NH2)2.xH2O Formula Weight: 251.11(anhy) Physical Form: Powder Odor: Odorless MDL No.: MFCD00049877

Formula: CoH₄N₂O₆S₂
Storage Temperature: Ambient
MDL Number: MFCD00049877
CAS Number: 14017-41-5
EINECS: 237-834-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR