Platinum 5% on activated carbon, powder, sulfided 0.5% S (as sulfide)
Supplier: Thermo Scientific Chemicals
|
Warning
|
43905-09
43905-18
CAAA43905-09EA
289.14
CAD
CAAA43905-09
CAAA43905-18
Platinum 5% on activated carbon, powder, sulfided 0.5% S (as sulfide)
Platinum
CAS No.: 7440-06-4
Physical Form: Powder
UN No.: 1325
MDL No.: MFCD00011179
Formula:
Pt Storage Temperature: Ambient |
MDL Number:
MFCD00011179 CAS Number: 7440-06-4 UN: 1325 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...