Tetraammineplatinum(II) nitrate (≥50% Pt) ≥99.99% (metals basis), Premion®
Supplier: Thermo Scientific Chemicals
|
Danger
|
43897-03
43897-06
CAAA43897-03EA
391.84
CAD
CAAA43897-03
CAAA43897-06
Tetraammineplatinum(II) nitrate (≥50% Pt) ≥99.99% (metals basis), Premion®
Tetraammineplatinum(II) nitrate
Purity: 99.99%
CAS No.: 20634-12-2
Molecular Formula: H12N6O6Pt
Formula Weight: 387.21
Physical Form: Crystalline
Appearance: White
Melting Point: 262° dec.
UN No.: 1477
MDL No.: MFCD00011624
Formula:
H₁₂N₆O₆Pt Melting Pt: 262 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00011624 CAS Number: 20634-12-2 EINECS: 243-929-9 UN: 1477 ADR: 5.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...