Polyethyleneimine, M.W. 25,000, linear
Supplier: Thermo Scientific Chemicals
|
Warning
|
43896-01
43896-03
CAAAA43896-01EA
1075.98
CAD
CAAAA43896-01
CAAA43896-03
Polyethyleneimine, M.W. 25,000, linear
Polyethyleneimine
CAS No.: 9002-98-6
Molecular Formula: -HN-[CH2CH2NH-]n
Physical Form: Powder
Odor: Amine-like
Melting Point: 59-60°
MDL No.: MFCD00084427
Formula:
(CH2CH2NH)n Melting Pt: 59…60 °C Storage Temperature: Ambient |
MDL Number:
MFCD00084427 CAS Number: 9002-98-6 |
Specification Test Results
Form | Powder |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...