Glycerol monostearate, purified
Supplier: Thermo Scientific Chemicals
43883-30
43883-A1
43883-A7
CAAA43883-30EA
69.84
CAD
CAAA43883-30
CAAA43883-A1
CAAA43883-A7
Glycerol monostearate, purified
Glycerol monostearate
Soluble in hot organic solvents
CAS No.: 31566-31-1
Molecular Formula: C17H35OCOCH2CH(OH)CH2OH
Formula Weight: 358.56
Physical Form: Powder
Solubility: Soluble in hot organic solvents
Merck Reference: 14,4489
MDL No.: MFCD00036186
Formula:
C₂₁H₄₄O₅ Storage Temperature: Ambient |
MDL Number:
MFCD00036186 CAS Number: 31566-31-1 EINECS: 250-705-4 Merck Index: 14,4489 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...