Sulfobromophthalein sodium salt hydrate
Supplier: Thermo Scientific Chemicals
43878-03
43878-06
43878-14
CAAA43878-03EA
56.25
CAD
CAAA43878-03
CAAA43878-06
CAAA43878-14
Sulfobromophthalein sodium salt hydrate
Sulfobromophthalein sodium salt hydrate
CAS No.: 71-67-0
Molecular Formula: C20H8Br4O10S2Na2.xH2O
Formula Weight: 837.98(anhy)
Physical Form: Powder
Sensitivity: Moisture Sensitive
Appearance: White
MDL No.: MFCD00150017
Formula:
C₂₀H₈Br₄O₁₀S₂Na₂·xH₂O Storage Temperature: Ambient |
MDL Number:
MFCD00150017 CAS Number: 71-67-0 EINECS: 200-761-0 Merck Index: 14,8954 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...