Platinum 10% (nominal) on carbon black, HiSPEC® 2000

Supplier: Thermo Scientific Chemicals
Danger

43876-02 43876-04 43876-09
CAAA43876-02EA 148.84 CAD
CAAA43876-02 CAAA43876-04 CAAA43876-09
Platinum 10% (nominal) on carbon black, HiSPEC® 2000
Platinum

CAS No.: 7440-06-4 Odor: Odorless UN No.: 1325 MDL No.: MFCD00011179

Formula: Pt
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
UN: 1325
ADR: 4.1,II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR