Fullerene nanotube multi-walled 20 nm OD, Length: 5-20 micron
Supplier: Thermo Scientific Chemicals
|
Warning
|
43839-MD
43839-MF
CAAA43839-MDEA
178.54
CAD
CAAA43839-MD
CAAA43839-MF
Fullerene nanotube multi-walled 20 nm OD, Length: 5-20 micron
Fullerene nanotube
Contains 95% nanotubes (TEM)
CAS No.: 308068-56-6
Physical Form: -270 mesh powder
Appearance: Black
UN No.: 1325
MDL No.: MFCD00133992
Storage Temperature:
Ambient |
MDL Number:
MFCD00133992 CAS Number: 308068-56-6 UN: 1325 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...