3-Methyl-1-phenylindolin-2-one 97

Supplier: Thermo Scientific Chemicals

43808-03 43808-06
CAAA43808-03EA 161.16 CAD
CAAA43808-03 CAAA43808-06
3-Methyl-1-phenylindolin-2-one 97
3-Methyl-1-phenylindolin-2-one

Purity: 97% CAS No.: 23210-22-2 Molecular Formula: C15H13NO Formula Weight: 223.27 Physical Form: Powder MDL No.: MFCD00798609

Formula: C₁₅H₁₃NO
MW: 223.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00798609
CAS Number: 23210-22-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR