2-Chloro-2-methylnonane 90+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
43784-06
43784-14
CAAA43784-06EA
129.56
CAD
CAAA43784-06
CAAA43784-14
2-Chloro-2-methylnonane 90+%
2-Chloro-2-methylnonane
Purity: 90+%
CAS No.: 4325-50-2
Molecular Formula: CH3(CH2)6C(CH3)(Cl)CH3
Formula Weight: 176.73
Physical Form: Liquid
Appearance: Colorless
UN No.: 3082
MDL No.: MFCD00060781
Formula:
C₁₀H₂₁Cl Storage Temperature: Ambient |
MDL Number:
MFCD00060781 CAS Number: 4325-50-2 UN: 3082 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...