Polydimethylsiloxane, hydroxy terminated, M.W. 4200

Supplier: Thermo Scientific Chemicals

Synonyms: Polysiloxanes, di-Me, hydroxy-terminated

43769-AD 43769-AK 43769-K2 43769-K4
CAAA43769-ADEA 50.24 CAD
CAAA43769-AD CAAA43769-AK CAAA43769-K2 CAAA43769-K4
Polydimethylsiloxane, hydroxy terminated, M.W. 4200
Polydimethylsiloxane, hydroxy terminated

Viscosity 90-120 centistokes


CAS No.: 70131-67-8 Molecular Formula: HO[-Si(CH3)2O-]nH Physical Form: Liquid Density: 0.97 MDL No.: MFCD01325012

Formula: [Si(CH₃)₂O]n
MW: 45.092 g/mol
Density: 0.97
Storage Temperature: Ambient
MDL Number: MFCD01325012
CAS Number: 70131-67-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR