Trimethyl(pentamethylcyclopentadienyl)titanium(IV) ≥97%

Supplier: Thermo Scientific Chemicals
Danger

43759-02 43759-04
CAAA43759-02EA 319.16 CAD
CAAA43759-02 CAAA43759-04
Trimethyl(pentamethylcyclopentadienyl)titanium(IV) ≥97%
Trimethyl(pentamethylcyclopentadienyl)titanium(IV)

Purity: 97% CAS No.: 107333-47-1 Molecular Formula: C13H24Ti Formula Weight: 228.22 Physical Form: Crystalline, Ampouled under argon Sensitivity: Light, Air and Moisture Sensitive UN No.: 1325 MDL No.: MFCD00269851

Formula: C₁₃H₂₄Ti
Storage Temperature: Ambient
MDL Number: MFCD00269851
CAS Number: 107333-47-1
UN: 1325
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR