Leuco malachite green
Supplier: Thermo Scientific Chemicals
43758-04
43758-09
CAAA43758-04EA
39.82
CAD
CAAA43758-04
CAAA43758-09
Leuco malachite green
Leuco malachite green
CAS No.: 129-73-7
Molecular Formula: C23H26N2
Formula Weight: 330.47
Physical Form: Powder
Sensitivity: Air Sensitive
Melting Point: 100-102°
Beilstein Reference: 2140506
MDL No.: MFCD00008315
Formula:
C₂₃H₂₆N₂ Melting Pt: 100…102 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008315 CAS Number: 129-73-7 EINECS: 204-961-9 Merck Index: 14,5699 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...