Fluorescamine

Supplier: Thermo Scientific Chemicals

43749-MB 43749-MC 43749-ME
CAAA43749-MBEA 58.14 CAD
CAAA43749-MB CAAA43749-MC CAAA43749-ME
Fluorescamine
Fluorescamine

For fluorescent detection of amino acids, peptides and proteins


Applications: For fluorescent detection of amino acids, peptides and proteins


Applications: For fluorescent detection of amino acids, peptides and proteins

Formula: C₁₇H₁₀O₄
Melting Pt: 155…158 °C
Storage Temperature: Ambient
MDL Number: MFCD00005928
CAS Number: 38183-12-9
EINECS: 253-814-5
Merck Index: 14,4158

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR