Crystal violet lactone

Supplier: Thermo Scientific Chemicals
Warning

43710-06 43710-14
CAAA43710-06EA 87.37 CAD
CAAA43710-06 CAAA43710-14
Crystal violet lactone
Crystal violet lactone

CAS No.: 1552-42-7 Molecular Formula: C26H29N3O2 Formula Weight: 415.53 Physical Form: Powder Melting Point: 180°-183° MDL No.: MFCD00070611

Formula: C₂₆H₂₉N₃O₂
Melting Pt: 180 °C…183 °C
Storage Temperature: Ambient
MDL Number: MFCD00070611
CAS Number: 1552-42-7
EINECS: 216-293-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR