Polyaniline, emeraldine salt from p-Toluenesulfonic acid

Supplier: Thermo Scientific Chemicals

Synonyms: Polyaniline ES, PAni ES

43683-09 43683-18
CAAA43683-09EA 178.54 CAD
CAAA43683-09 CAAA43683-18
Polyaniline, emeraldine salt from p-Toluenesulfonic acid
Polyaniline, emeraldine salt

Applications: Polyaniline is used in chemical vapor sensors, supercapacitors and biosensors. It is also useful for manufacture of electrical conducting yarns, antistatic coatings, electromagnetic shielding, flexible electrodes and printed circuit board. Further, it is used in antistatic and ESD coatings and corrosion protection.


CAS No.: 25233-30-1 Physical Form: Powder Appearance: Dark green Melting Point: 300° dec. MDL No.: MFCD00284320

Formula: C₃H₅BrClN₃
Melting Pt: >300 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00284320
CAS Number: 25233-30-1

Order Now

Specification Test Results

Form Powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR