Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 410
Supplier: Thermo Scientific Chemicals
Warning
|
43669-AD
43669-AK
43669-K2
CAAA43669-ADEA
94.01
CAD
CAAA43669-AD
CAAA43669-AK
CAAA43669-K2
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 410
Polydimethylsiloxane, trimethylsiloxy terminated
Viscosity 2.0 centistokes
CAS No.: 9016-00-6
Molecular Formula: (CH3)3SiO(Si(CH3)2O))nSi(CH3)3
Flash Point: 70°(158°F)
Physical Form: Liquid
Density: 0.88
Refractive Index: 1.3900
Appearance: Colorless
Odor: Odorless
Melting Point: -40°
Boiling Point: 85-105°/2-4mm
MDL No.: MFCD00148360
Formula:
(CH₃)₃SiO(Si(CH₃)₂O)nSi(CH₃)₃ Boiling Pt: 85…105 °C (2…4 mmHg) Melting Pt: –40 °C Density: 0.88 Flash Pt: 70 °C (158 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00148360 CAS Number: 9016-00-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...