4-(2-Pyridylazo)resorcinol ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
43629-03
43629-06
CAAA43629-03EA
51.67
CAD
CAAA43629-03
CAAA43629-06
4-(2-Pyridylazo)resorcinol ACS
4-(2-Pyridylazo)resorcinol
CAS No.: 1141-59-9
Molecular Formula: C11H9N3O2
Formula Weight: 215.21
Physical Form: Powder
Melting Point: 187° dec.
Beilstein Reference: 184665
Formula:
C₁₁H₉N₃O₂ Melting Pt: 187 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00044237 CAS Number: 1141-59-9 EINECS: 214-528-6 |
Specification Test Results
Clarity of solution | Pass test |
Suitability as a complexometric ion indicator | Pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...