Etidronic acid ≥96%

Supplier: Thermo Scientific Chemicals
Danger

43502-18 43502-09
CAAAA43502-18EA 737.86 CAD
CAAAA43502-18 CAAA43502-09
Etidronic acid ≥96%
Etidronic acid

Purity: 96% CAS No.: 2809-21-4 Molecular Formula: CH3C(OH)(PO3H2)2 Formula Weight: 206.03 Physical Form: Powder Odor: Odorless Merck Reference: 14,3863 UN No.: 3261 MDL No.: MFCD00070585

Formula: C₂H₈O₇P₂
Storage Temperature: Ambient
MDL Number: MFCD00070585
CAS Number: 2809-21-4
EINECS: 220-552-8
UN: 3261
ADR: 8,III
Merck Index: 14,3863

Order Now

Specification Test Results

Form Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR