Ammonium tetrathiomolybdate ≥99.95% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
43493-06
43493-14
CAAA43493-06EA
202.24
CAD
CAAA43493-06
CAAA43493-14
Ammonium tetrathiomolybdate ≥99.95% (metals basis)
Ammonium tetrathiomolybdate
Purity: 99.95%
CAS No.: 15060-55-6
Molecular Formula: H8MoN2S4
Formula Weight: 260.27
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: Black
MDL No.: MFCD00136013
Formula:
H₁₀MoN₂S₄ Storage Temperature: Ambient |
MDL Number:
MFCD00136013 CAS Number: 15060-55-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...