Gallium trinitrate solution (9 - 10% Ga) (w/w)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Gallium (III) nitrate

43484-18 43484-30
CAAA43484-18EA 208.56 CAD
CAAA43484-18 CAAA43484-30
Gallium trinitrate solution (9 - 10% Ga) (w/w)
Gallium trinitrate

contains 25-30% HNO₃


Purity: 9-10% CAS No.: 13494-90-1 Molecular Formula: Ga(NO3)3 Formula Weight: 255.73 Physical Form: Liquid UN No.: 3218 MDL No.: MFCD00011016

Formula: GaN₃O₉
MW: 255.74
Melting Pt: 110
Storage Temperature: Ambient
MDL Number: MFCD00011016
CAS Number: 13494-90-1
EINECS: 236-815-5
UN: 3218
ADR: 5.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR