6-Methyl-4-trifluoromethyl-2(1H)-pyridone ≥97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Methyl-4-(trifluoromethyl)pyridin-2(1H)-one

43467-03 43467-06
CAAAA43467-01EA 78.51 CAD
CAAAA43467-01 CAAA43467-03 CAAA43467-06 CAAA43467-14
6-Methyl-4-trifluoromethyl-2(1H)-pyridone ≥97%
6-Methyl-4-trifluoromethyl-2(1H)-pyridone

Purity: 97% CAS No.: 22123-19-9 Molecular Formula: C7H6F3NO Formula Weight: 177.13 Physical Form: Powder Melting Point: 129-130° MDL No.: MFCD00042479

Formula: C₇H₆F₃NO
MW: 177.126 g/mol
Melting Pt: 129…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00042479
CAS Number: 22123-19-9

Order Now

Specification Test Results

Form Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR