Stainless steel (Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%), powder -10+20 mesh Type 316-L
Supplier: Thermo Scientific Chemicals
Danger
43458-36 43458-A3
CAAA43458-36EA 133.19 CAD
CAAA43458-36 CAAA43458-A3
Stainless steel (Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%), powder -10+20 mesh Type 316-L
Stainless steel
Molecular Formula: Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt% Density: 1.57 Appearance: Grey MDL No.: MFCD00148499