Ruthenium(III) nitrosylsulfate ≥99.99% (metals basis), Premion®
Supplier: Thermo Scientific Chemicals
|
Warning
|
43436-03
43436-06
CAAA43436-03EA
161.16
CAD
CAAA43436-03
CAAA43436-06
Ruthenium(III) nitrosylsulfate ≥99.99% (metals basis), Premion®
Ruthenium(III) nitrosylsulfate
Applications: Electroplating, coating
Soluble in water
Purity: 99.99%
Molecular Formula: [Ru(NO)]2(SO4)3
Formula Weight: 550.34
Physical Form: Powder
Solubility: Soluble in water
MDL No.: MFCD02684508
Applications: Electroplating, coating
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...