Ruthenium(III) nitrosylacetate ≥99.99% (metals basis), Premion®
Supplier: Thermo Scientific Chemicals
|
Warning
|
43435-03
43435-06
CAAA43435-03EA
199.08
CAD
CAAA43435-03
CAAA43435-06
Ruthenium(III) nitrosylacetate ≥99.99% (metals basis), Premion®
Ruthenium(III) nitrosylacetate
Purity: 99.99%
Molecular Formula: Ru(NO)(OOCCH3)3
Formula Weight: 308.18
Physical Form: Powder
MDL No.: MFCD02684507
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...