4-Bromomethyl-6,7-dimethoxycoumarin ≥95%

Supplier: Thermo Scientific Chemicals
Danger

43402-04 43402-77
CAAAA43402-04EA 614.62 CAD
CAAAA43402-04 CAAAA43402-77
4-Bromomethyl-6,7-dimethoxycoumarin ≥95%
4-Bromomethyl-6,7-dimethoxycoumarin

Applications: Fluorescent labeling reagent. used in HPLC for the detection of carboxylic acids


Applications: Fluorescent labeling reagent. Used in HPLC for the detection of carboxylic acids


Applications: Fluorescent labeling reagent. Used in HPLC for the detection of carboxylic acids

Formula: C₁₂H₁₁BrO₄
Melting Pt: 200…203 °C
Storage Temperature: Ambient
MDL Number: MFCD00011570
CAS Number: 88404-25-5
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Form Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR