Methyl-4-aminothiophene-3-carboxylate hydrochloride ≥97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
43384-03
43384-06
CAAA43384-03EA
138.57
CAD
CAAA43384-03
CAAA43384-06
Methyl-4-aminothiophene-3-carboxylate hydrochloride ≥97%
Methyl-4-aminothiophene-3-carboxylate hydrochloride
Purity: 97+%
CAS No.: 39978-14-8
Molecular Formula: C6H7NO2S.HCl
Formula Weight: 193.65
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: Brown
Melting Point: 199-200° dec.
MDL No.: MFCD00068149
Formula:
C₆H₈ClNO₂S Melting Pt: 199…200 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00068149 CAS Number: 39978-14-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...