Paraformaldehyde 16% (w/v) in aqueous solution methanol-free

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde

43368-9L 43368-9M
CAAA43368-9LEA 76.79 CAD
CAAA43368-9L CAAA43368-9M
Paraformaldehyde 16% (w/v) in aqueous solution methanol-free
Formaldehyde

Solution is micro-filtered and supplied in ampules. RNase and DNase free.


CAS No.: 30525-89-4 Molecular Formula: (CH2O)n Physical Form: Liquid, Ampouled under argon MDL No.: MFCD00133991

Formula: CH₂O
MW: 30.03 g/mol
Boiling Pt: -19.3
Melting Pt: -92
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
UN: 3334
ADR: 9 III
Merck Index: 13,04259

Order Now

Specification Test Results

Concentration (w/v) 15.5 to 16.5%
Appearance Clear colourless liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR