Yttrium(III) 2-ethylhexanoate 10% (w/v) in n-hexane

Supplier: Thermo Scientific Chemicals
Danger

43356-AC 43356-AE
CAAA43356-ACEA 85.95 CAD
CAAA43356-AC CAAA43356-AE CAAA43356-AP
Yttrium(III) 2-ethylhexanoate 10% (w/v) in n-hexane
Yttrium (III) 2-ethylhexanoate

CAS No.: 103470-68-4 Molecular Formula: C24H45O6Y Formula Weight: 518.16 Physical Form: Liquid UN No.: 1208 MDL No.: MFCD00070441

Formula: C₂₄H₄₅O₆Y
MW: 518.524 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070441
CAS Number: 103470-68-4
UN: 1208
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR