Polyethyleneimine, beads_silica 20-40 mesh exchange resin_anion
Supplier: Thermo Scientific Chemicals
Synonyms:
Polyaziridine, Corcat, Polyethylenimine, Polymin P, Polyaziridine Mw 25000, Aziridine, PEI, Polyethylenimine (branched), Ethyleneimine Polymer, Poly(ethyleneimine)
Anion exchange resin for the separation of nucleotide mixtures
These resins, though solid, are sold by volume since exchange capacity is related to volume
CAS No.: 9002-98-6
Physical Form: Beads
Sensitivity: Hygroscopic
MDL No.: MFCD00084427
Applications: For the separation of nucleotide mixtures
Formula:
(CH2CH2NH)n Storage Temperature: Ambient |
MDL Number:
MFCD00084427 CAS Number: 9002-98-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...