3-(Trifluoromethyl)phenyltrimethylammonium hydroxide 5% (w/v) in methanol

Supplier: Thermo Scientific Chemicals
Danger

43284-AA 43284-AC
CAAA43284-AAEA 191.18 CAD
CAAA43284-AA CAAA43284-AC
3-(Trifluoromethyl)phenyltrimethylammonium hydroxide 5% (w/v) in methanol
3-(Trifluoromethyl)phenyltrimethylammonium hydroxide

Applications: Transesterification reactions


Applications: Transesterification reactions


Applications: Transesterification reactions

Formula: C₁₀H₁₄F₃NO
Storage Temperature: Ambient
MDL Number: MFCD00059473
CAS Number: 68254-41-1
EINECS: 200-659-6
UN: 3286
ADR: 3,II

Order Now

Specification Test Results

Form Liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR