Zirconium carbonate basic hydrate ≥97%

Supplier: Thermo Scientific Chemicals

43245-36 43245-A3
CAAA43245-36EA 161.16 CAD
CAAA43245-36 CAAA43245-A3
Zirconium carbonate basic hydrate ≥97%
Zirconium carbonate basic hydrate

CAS No.: 57219-64-4 Molecular Formula: [ZrO2]2.CO2.xH2O Formula Weight: 290.45(anhy) Physical Form: Powder Density: 2.2 Appearance: White MDL No.: MFCD00172265

Formula: C₄H₄O₁₂Zr
Density: 2.2
Storage Temperature: Ambient
MDL Number: MFCD00172265
CAS Number: 57219-64-4
EINECS: 260-633-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR