DMS (Dimethyl suberimidate dihydrochloride)

Supplier: Thermo Scientific Chemicals
Warning

43244-04 43244-09
CAAA43244-04EA 95.91 CAD
CAAA43244-04 CAAA43244-09
DMS (Dimethyl suberimidate dihydrochloride)
DMS (Dimethyl suberimidate dihydrochloride)

Applications: Crosslinking of proteins and poly(allylamine)
Soluble in water


CAS No.: 34490-86-3 Molecular Formula: C10H20N2O2.2HCl Formula Weight: 273.20 Physical Form: Crystalline Sensitivity: Moisture Sensitive Melting Point: 213-214° Solubility: Soluble in water MDL No.: MFCD00012574


Applications: Crosslinking of proteins and poly(allylamine)

Formula: C₁₀H₂₀N₂O₂
Melting Pt: 213…214 °C
Storage Temperature: Ambient
MDL Number: MFCD00012574
CAS Number: 34490-86-3
EINECS: 252-060-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR