Tris(dimethylamino)aluminum dimer
Supplier: Thermo Scientific Chemicals
|
Danger
|
43237-03
43237-06
CAAA43237-03EA
274.92
CAD
CAAA43237-03
CAAA43237-06
Tris(dimethylamino)aluminum dimer
Tris(dimethylamino)aluminum dimer
Applications: Preparation of aluminum nitrides
CAS No.: 32093-39-3
Molecular Formula: C12H36Al2N6
Formula Weight: 318.42
Flash Point: 21°(70°F)
Physical Form: Powder, packaged under argon
Density: 0.865
Sensitivity: Moisture Sensitive
Appearance: White to pale brown
Melting Point: 87-89°
Boiling Point: 90°/0.05mm subl.
UN No.: 3131
MDL No.: MFCD00269811
Applications: Preparation of aluminum nitrides
Formula:
C₁₂H₃₆Al₂N₆ Boiling Pt: 90 °C (0.05 mmHg) (subl.) Melting Pt: 87…89 °C Density: 0,865 Flash Pt: 21 °C (70 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00269811 CAS Number: 32093-39-3 UN: 3131 ADR: 4.3,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...