Poly(methyl-3,3,3-trifluoropropylsiloxane), M.W. 14,000

Supplier: Thermo Scientific Chemicals
Warning

43230-14 43230-22
CAAA43230-06EA 50.4 CAD
CAAA43230-06 CAAA43230-14 CAAA43230-22
Poly(methyl-3,3,3-trifluoropropylsiloxane), M.W. 14,000
Poly(methyl-3,3,3-trifluoropropylsiloxane)

Uses: Fluorosilicone lubricant
Viscosity 10,000 centistokes


CAS No.: 63148-56-1 Molecular Formula: [-Si(CH3)(CH3CH2CF3)O-]n Flash Point: 315°(599°F) Physical Form: Liquid Density: 1.30 Refractive Index: 1.383 MDL No.: MFCD00241477


Applications: Fluorosilicone lubricant

Formula: [-Si(CH₃)(CH₃CH₂CF₃)O-]n
Density: 1,30
Flash Pt: 315 °C (599 °F)
Storage Temperature: Ambient
MDL Number: MFCD00241477
CAS Number: 63148-56-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR