Poly(methyl-3,3,3-trifluoropropylsiloxane), M.W. 2,400
Supplier: Thermo Scientific Chemicals
|
Warning
|
43228-14
43228-22
CAAA43228-06EA
39.12
CAD
CAAA43228-06
CAAA43228-14
CAAA43228-22
Poly(methyl-3,3,3-trifluoropropylsiloxane), M.W. 2,400
Poly(methyl-3,3,3-trifluoropropylsiloxane)
Uses: Fluorosilicone lubricant
Viscosity 300-350 centistokes
CAS No.: 63148-56-1
Molecular Formula: [-Si(CH3)(CH3CH2CF3)O-]n
Flash Point: 260°(500°F)
Physical Form: Liquid
Density: 1.25
Refractive Index: 1.381
Appearance: Colorless
MDL No.: MFCD00241477
Applications: Fluorosilicone lubricant
Formula:
[-Si(CH₃)(CH₃CH₂CF₃)O-]n Density: 1.25 Flash Pt: 260 °C (500 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00241477 CAS Number: 63148-56-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...