Graphite, nickel coated (≥99.8% Ni) (metals basis), powder -100 mesh
Supplier: Thermo Scientific Chemicals
|
Danger
|
43215-22
43215-36
CAAA43215-22EA
70.47
CAD
CAAA43215-22
CAAA43215-36
Graphite, nickel coated (≥99.8% Ni) (metals basis), powder -100 mesh
Graphite, nickel coated
Applications: Conductive surface coatings, inks, adhesives, and sealants
Purity: 99.8%
Physical Form: Ni:C; 60:40 wt%
Appearance: Black
Odor: Odorless
UN No.: 3178
MDL No.: MFCD00144065
Applications: Conductive surface coatings, inks, adhesives, and sealants
Storage Temperature:
Ambient |
MDL Number:
MFCD00144065 UN: 3178 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...