Zirconium disulfate tetrahydrate ≥99.99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
43194-14
43194-22
CAAA43194-14EA
281.24
CAD
CAAA43194-14
CAAA43194-22
Zirconium disulfate tetrahydrate ≥99.99% (metals basis)
Zirconium disulphate tetrahydrate
Purity: 99.99%
CAS No.: 7446-31-3
Molecular Formula: Zr(SO4)2.4H2O
Formula Weight: 355.40 (283.34anhy)
Physical Form: Crystalline powder
Density: 3.22
Appearance: White
Merck Reference: 14,10182
MDL No.: MFCD00149267
Formula:
O₈S₂Zr·4H₂O Density: 3.22 Storage Temperature: Ambient |
MDL Number:
MFCD00149267 CAS Number: 7446-31-3 EINECS: 238-694-4 Merck Index: 14,10182 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...