Lithium niobium isopropoxide 5% (w/v) in isopropanol (IPA) , 99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
43175-AC
43175-AE
CAAA43175-ACEA
145.2
CAD
CAAA43175-AC
CAAA43175-AE
Lithium niobium isopropoxide 5% (w/v) in isopropanol (IPA) , 99% (metals basis)
Lithium niobium isopropoxide
Purity: 99+%
CAS No.: 21864-21-1
Molecular Formula: C18H42LiNbO6
Formula Weight: 454.38
Physical Form: Liquid
Sensitivity: Moisture Sensitive
Odor: Alcohol-like
UN No.: 1219
MDL No.: MFCD00210626
Formula:
LiNb(OC₃H₇)₆ Storage Temperature: Ambient |
MDL Number:
MFCD00210626 CAS Number: 21864-21-1 UN: 1219 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...