Palladium 5% on calcium carbonate Catalyst lead poisoned Type A306060-5

Supplier: Thermo Scientific Chemicals
Danger

43172-06 43172-14 43172-22
CAAA43172-06EA 130.67 CAD
CAAA43172-06 CAAA43172-14 CAAA43172-22
Palladium 5% on calcium carbonate Catalyst lead poisoned Type A306060-5
Palladium

Applications: Selective hydrogenation


CAS No.: 7440-05-3 Physical Form: Powder UN No.: 3077 MDL No.: MFCD03427452


Applications: Selective hydrogenation

Formula: Pd
MW: 106.420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
UN: 3077
ADR: 9 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR