Lanthanum(III) phosphate hydrate

Supplier: Thermo Scientific Chemicals

43125-14 43125-22
CAAA43125-14EA 116.45 CAD
CAAA43125-14 CAAA43125-22
Lanthanum(III) phosphate hydrate
Lanthanum(III) phosphate hydrate

CAS No.: 14913-14-5 Molecular Formula: LaPO4.xH2O Formula Weight: 233.88(anhy) Physical Form: Lump Odor: Odorless MDL No.: MFCD00150248

Formula: LaPO₄.xH₂O
MW: 233.877 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00150248
CAS Number: 14913-14-5
EINECS: 237-419-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR